ESD safety shoes, sandals, Velcro® fastening, Scott
Supplier: ELTEN
Certificates
About this item
100% metal- and leather-free, lightweight ESD safety sandal made from microfibre and textile upper, reflective materials, a padded tongue plus a slip resistant, dual-density TPU/PU sole (Sportics).
- Climate-regulating, breathable textile lining
- Full length inlay sole (ESD Pro), black
- Soft fleece ESD insole
- Composite toe protection cap (200 J)
Ideal for use in dry work areas such as industry, storage, transport, or assembly.
Colour: Black
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S1, SRC.
Specifications
- Catalog No:
- ELTN71872/42
- ELTN71872/37
- ELTN71872/38
- ELTN71872/35
- ELTN71872/47
- ELTN71872/36
- ELTN71872/48
- ELTN71872/45
- ELTN71872/46
- ELTN71872/43
- ELTN71872/44
- ELTN71872/40
- ELTN71872/41
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001