Order Entry
Austria
ContactUsLinkComponent
VWR® pHenomenal® MD 8000 L Bench Top Digital Multi-Parameter Instruments
VWR® pHenomenal® MD 8000 L Bench Top Digital Multi-Parameter Instruments
Supplier:  VWR Collection
Certificates
undefined
VWR® pHenomenal® MD 8000 L Bench Top Digital Multi-Parameter Instruments
Supplier:  VWR Collection

Specifications

  • Catalog No:
  • 665-0499
  • 665-0498
  • pH range:
  • 0 - 14
  • 0 - 14
  • pH resolution:
  • 0,1/0,01/0,001
  • 0,1/0,01/0,001
  • pH accuracy:
  • ±0,1; ±0,01; ±0,005
  • ±0,1; ±0,01; ±0,005
  • mV range:
  • ±2500; ±1200
  • ±2500; ±1200
  • mV resolution:
  • 0,1; 1
  • 0,1; 1
  • mV accuracy:
  • ±0,3; ±1
  • ±0,3; ±1
  • Conductivity range:
  • 0,01 µS/cm - 1000 mS/cm (depending on sensor)
  • 0,01 µS/cm - 1000 mS/cm (depending on sensor)
  • Conductivity accuracy:
  • ±0,5% of value
  • ±0,5% of value
  • Conductivity cell constant:
  • Automatic
  • Automatic
  • Resistivity range:
  • 1 Ωcm - 20 MΩcm (depending on sensor)
  • 1 Ωcm - 20 MΩcm (depending on sensor)
  • Salinity range:
  • 0 - 70
  • 0 - 70
  • TDS range:
  • 0 mg/L - 199,9 g/L
  • 0 mg/L - 199,9 g/L
  • DO concentration:
  • 0,00 - 20,00 / 0,0 - 50,0 mg/L
  • 0,00 - 20,00 / 0,0 - 50,0 mg/L
  • DO saturation:
  • 0,0 - 200,0; 0 - 500 %
  • 0,0 - 200,0; 0 - 500 %
  • Temperature range:
  • Auto: −5...+105 °C
  • Auto: −5...+105 °C
  • Temperature resolution:
  • 0,1 °C
  • 0,1 °C
  • Temperature accuracy:
  • ±0,2 K
  • ±0,2 K
  • Display:
  • Blacklit B/W graphic
  • Blacklit B/W graphic
  • Power supply:
  • 230 V with power supply or 4× 1,5 V AA (supplied) or via USB port
  • 230 V with power supply or 4× 1,5 V AA (supplied) or via USB port
  • Data logging - log function:
  • Yes
  • Yes
  • Data logging capacity:
  • Up to 4500 data sets in automatic
  • Up to 4500 data sets in automatic
  • IP rating:
  • IP 43
  • IP 43
  • Connections:
  • 1 measurement input
  • 1 measurement input
  • Reference temperature:
  • 20 / 25 °C
  • 20 / 25 °C
  • Interfaces:
  • Mini USB-B (USB 1.1) for data downloads
  • Mini USB-B (USB 1.1) for data downloads
  • Parameters:
  • pH, mV, temperature, conductivity, dissolved oxygen, turbidity
  • pH, mV, temperature, conductivity, dissolved oxygen, turbidity
  • Calibration:
  • Anycal 5-point customer buffer calibration
    Max. 10 calibration storage
    1 - 999 days calibration timer
  • Anycal 5-point customer buffer calibration
    Max. 10 calibration storage
    1 - 999 days calibration timer
  • Calibration points:
  • 1 - 5 (automatic buffer recognition)
  • 1 - 5 (automatic buffer recognition)

Specifications

pH electrode

Product Information

Frequently Bought Together