Lactalbumin hydrolysate
1.12523.5000
: Merck
:
- Pk:5 kg
- Seq:34
- Protein/peptide name:Lactalbumin hydrolysate
CAS No.: 68458-87-7
Physical Form: Powder
Appearance: Tan
MDL No.: MFCD00147378
Applications: A solubilized enzymatic digest of lactalbumin