Order Entry
Austria
ContactUsLinkComponent
Anti-SYUA Chicken Polyclonal Antibody
Anti-SYUA Chicken Polyclonal Antibody
Catalog # BSENC-1795-50
Supplier:  Biosensis
undefined
Anti-SYUA Chicken Polyclonal Antibody
Catalog # BSENC-1795-50
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Alpha-synuclein
  • Antigen symbol:
    SYUA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Chicken
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Pig,
    Horse,
    Chicken,
    Human,
    Bovine,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Format:
    Purified IgY
  • Form:
    Lyophilized
  • Antigen synonyms:
    Non-A beta component of AD amyloid|Non-A4 component of amyloid precursor|NACP
  • Amino acid number:
    61 to 95
  • UniProtKB:
    Uniprot # P37840
  • Storage temperature:
    Store lyophilised antibody at 2-8 °C. After reconstitution divide into aliquots and store at –20 °C for long-term storage. Store at 2-8 °C short-term (up to 4 weeks). Avoid repetitive freeze/thaw cycles.
  • Shipping temperature:
    Ambient
  • Immunogen:
    Full length human protein with the epitope from amino acids 61-95
  • Purification:
    Affinity purified
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

May be involved in the regulation of dopamine release and transport. Induces fibrillization of microtubule-associated protein tau. Reduces neuronal responsiveness to various apoptotic stimuli, leading to a decreased caspase-3 activation. Ref: uniprot.org.

Western blotting (1:1,000 - 1:2,000), Immunocytochemistry (1:1,000 - 1:2,000) and Immunohistochemistry (1:1,000 1: 2,000). Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: SYUA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat, Cow, Pig, Horse, Chicken