Orders LinkContactUsLinkComponent
Utility washer
Utility washer
Utility washer for NMR tubes
Catalog # KLEI459960-0000
Supplier:   KLEINFELD
Utility washer
Catalog # KLEI459960-0000
Supplier:  KLEINFELD
Utility washer
Utility washer
Utility washer for NMR tubes
Price
592,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
This washer is used for rapid, safe washing and drying of cuvettes, tubes and small scale volumetric items up to approximately 22 mm OD.
  • Operates efficiently with a conventional sink aspirator
  • Recommended for 10 mm NMR tubes
  • Flushing is activated by lightly pressing the inverted cell onto a neoprene gasket
All glass apparatus manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery information: Supplied with one gasket, a tube washer, an Erlenmeyer flask and a ³/₈" hose connection.
Specifications
  • Description:
    Utility washer for NMR tubes