Order Entry
Austria
ContactUsLinkComponent
EDTA disodium salt 0.1 mol/l (0.2 N) in aqueous solution, AVS TITRINORM for hardness of water determinations
EDTA disodium salt 0.1 mol/l (0.2 N) in aqueous solution, AVS TITRINORM for hardness of water determinations
Catalog # VWRS28662.5000
Supplier:  VWR Collection
CAS Number:  
EDTA disodium salt 0.1 mol/l (0.2 N) in aqueous solution, AVS TITRINORM for hardness of water determinations
Catalog # VWRS28662.5000
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.0998 - 0.1002 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
 :336.21 g/mol
Storage Temperature:Ambient
EINECS:205-358-3
CAS Number:139-33-3
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672