

Formalin 10% stabilised, TECHNICAL, buffered (pH 7.0 ± 0.2)
Supplier: VWR Collection
Certificates
About this item
CAS Number:50-00-0
:
Formula:CH₂O
MDL Number:MFCD00003274
:Stabilised with methanol 0,5-1,5 %
:30.03 g/mol
EINECS:200-001-8
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (Formaldehyde):3.5 - 4.5 %
- pH (20°C):6.9 - 7.1
- Methanol:0.5 - 1.5 %
Specification Test Results
Related Information
Certificates
Search by Lot Numbersds_VWRS9713
SDS
spec_VELS9713.5001 spec_VWRS9713.0800 spec_VWRS9713.0800 spec_VWRS9713.0800 spec_VWRS9713.6937
Product specification
More
Frequently Bought Together