Order Entry
Austria
ContactUsLinkComponent
 
Dulbecco's phosphate buffered saline (DPBS), for cell culture, powder format, HyClone™
 
undefined
Dulbecco's phosphate buffered saline (DPBS), for cell culture, powder format, HyClone™
 

 

  • pH range:
    7,0 - 7,2
  • Storage temperature:
    2…8 °C

 

 

Frequently Bought Together