ContactUsLinkComponent
Electrolyte for reference electrodes
Electrolyte for reference electrodes
Catalog # METT51350088
Supplier:  Mettler - Toledo
CAS Number:  
Electrolyte for reference electrodes
Catalog # METT51350088
Supplier:  Mettler - Toledo
CAS Number:  
Specifications
  • Pk:
    6
  • Description:
    LiCl solution in 1 mol/l in ethanol, 6×30 ml
Specifications
About this item
Used for the measurement in non-aqueous media.