Order Entry
Austria
ContactUsLinkComponent
Cloth tape, Tesa®
Cloth tape, Tesa®
Catalog # KROS13493
Supplier:  KROSCHKE
Cloth tape, Tesa®
Catalog # KROS13493
Supplier:  KROSCHKE

Specifications

  • Width:
    50,0 mm
  • Description:
    Cloth tape
  • Pk:
    1

Specifications

About this item

Plastic-coated, strongly adhesive premium fabric tape, universally applicable.

  • Very easy to tear by hand
  • Writable
  • Resistant to aging, abrasion
  • Water- and weatherproof
  • Temperature resistant: 30 minutes at 130 °C

The tape should only be applied at normal room temperature.