Order Entry
Austria
ContactUsLinkComponent
 
Atto 647N-biotin, BioReagent
  SIAL93606-1MG
 :  Merck
 :  
Atto 647N-biotin, BioReagent
  SIAL93606-1MG
 :  Merck
 :  

 

  • Pk:
    1 mg
  • Description:
    Atto 647N-biotin

 

 

Designed for high sensitivity applications, including single molecule detection. These labels have rigid structures that do not show any cis-trans-isomerisation and display exceptional intensity with minimal spectral shift on conjugation.

  • Assay: ≥95% (HPCE)
  • Fluorescence: λex 644 nm; λem 660 nm in 0,1 M phosphate pH 7,0
  • Suitable for fluorescence