ContactUsLinkComponent
Protective hand creams, Physioderm®, SANSIBAL®
Protective hand creams, Physioderm®, SANSIBAL®
Catalog # FAWE13632-003
Supplier:  FAWECO HAUTSCHUTZ
undefined
Protective hand creams, Physioderm®, SANSIBAL®
Catalog # FAWE13632-003
Supplier:  FAWECO HAUTSCHUTZ
Specifications
  • Description:
    Protective hand cream, SANSIBAL®, 100 ml tube
  • Pk:
    12
Specifications
About this item
The water-soluble, pigment-based emulsion develops a separating and a protective film. This solid grid effectively prevents water-insoluble hazards from penetrating the upper layers of the skin. The protective film also reduces the adherence of dirt.
  • O/W emulsion
  • Slightly greasy cream
  • Silicone-free, perfumed
  • skin-neutral pH value
  • Fast absorbing
Suitable when handling water-insoluble substances, e.g. oil, grease or non-water-based cooling lubricants.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904