Optical magnifier
OPTK4987
: OPTIKA
- Magnification:8×
- Description:Optical magnifier
- Environmentally Preferable:
- Pk:1
- W×D:42×45 mm
This optical magnifier is designed with front opening and millimetric scale for accurate measurements.
- Focusing adjustment with 0,1 mm intervals
- Black linen scale design