ContactUsLinkComponent
Affinity chromatography media, HiTrap™ Capto™ Lentil Lectin
Affinity chromatography media, HiTrap™ Capto™ Lentil Lectin
Catalog # 17-5489-11
Supplier:  Cytiva
CAS Number:  
undefined
Affinity chromatography media, HiTrap™ Capto™ Lentil Lectin
Catalog # 17-5489-11
Supplier:  Cytiva
CAS Number:  
Specifications
  • Pk:
    5
  • Description:
    HiTrap™ Capto™ Lentil Lectin
  • Type:
    Liquid
Specifications
About this item
Capto™ Lentil Lectin is an affinity chromatography resin for purification of glycoproteins and other molecules containing carbohydrates such as α-D-mannose and α-D-glucose or sterically related residues.
  • Group-specific adsorbent for carbohydrate-containing molecules
  • High productivity and cost-efficiency in downstream operations
  • Animal component-free production
  • Security of supply and regulatory support
Capto™ Lentil Lectin resin is based on a rigid high-flow agarose base matrix that enables high flow rates. The product is available in bulk as well as in various high-throughput process development formats.