Orders LinkContactUsLinkComponent
Extraction unit, MSE29, Behrotest®
Extraction unit, MSE29, Behrotest®
Behrotest funnel separator, MSE 29
Catalog # BEHRB00217680
Supplier:   BEHR
Extraction unit, MSE29, Behrotest®
Catalog # BEHRB00217680
Supplier:  BEHR
Extraction unit, MSE29, Behrotest®
Behrotest funnel separator, MSE 29
Price
263,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Behrotest funnel separator is used for the determination of PAHs from water.
The design of Behrotest® funnel separator facilitates simple, clean separation of the phases or the emulsion. The entire organic phase can easily be transferred in the movable separator funnel top for further processing.
The Behrotest® funnel separator is used for the isolation of lipophilic substances from water using liquid phase extraction. The various functional groups of the system can be separated. It also makes easy to clean the individual functional groups.
Certifications: Conforms with DIN 38407 F18.
Specifications
  • Type:
    MSE 29
  • Description:
    Behrotest funnel separator