ContactUsLinkComponent
Mini mailbox P650 forwarding set white
Mini mailbox P650 forwarding set white
Catalog # DAKL520300
Supplier:  DAKLAPACK
undefined
Mini mailbox P650 forwarding set white
Catalog # DAKL520300
Supplier:  DAKLAPACK
Specifications
  • Description:
    Mini mailbox P650 forwarding set, white
  • W×D×H:
    240×129×30 mm
  • Pk:
    100
Specifications
About this item
The patented MiniMailBox system is approved for use in every country as a recognised safe way of transporting biological substances category B, UN3373 / P650.
  • Flap closure
  • Made from white cardboard
Commonly used system used by companies, hospitals and testing facilities.
Delivery information: The kit contains the MiniMailBox, Safetybag with bubble liner and absorbing 4 position tube holder.