ContactUsLinkComponent
Caps for autosampler vials
Caps for autosampler vials
Catalog # MICS97010-SC18
Supplier:  MICROSOLV
undefined
Caps for autosampler vials
Catalog # MICS97010-SC18
Supplier:  MICROSOLV
Specifications
  • Type:
    Polyethylene-push on
  • Description:
    Small caps, use with ND18 vials, 19,8×5,2 mm
  • Cap colour:
    Transparent
  • Pk:
    100