Order Entry
Austria
ContactUsLinkComponent
Storage vials, quick/snap
Storage vials, quick/snap
Catalog # MICS97010-CV05
Supplier:  MICROSOLV
undefined
Storage vials, quick/snap
Catalog # MICS97010-CV05
Supplier:  MICROSOLV

Specifications

  • Cap included:
    Yes
  • Ø×H:
    40×22 mm
  • Capacity:
    5,00 ml
  • Colour:
    Clear
  • Description:
    Vials, quick snap storage, ND18 snap
  • Pk:
    100

Specifications

About this item

Easy to use storage vials and caps with push on technology for ease and comfort.

  • Easy to handle
  • No liners required in the cap

To store samples that are not critical.

Caps

Product Information