Recombinant protein G, peroxidase conjugated, Calbiochem®
539322-500
:
- Pk:500 µG
- Description:Recombinant Protein G, peroxidase conjugated
Purified recombinant Protein G conjugated to peroxidase can be used in direct immunoassay.
- Used in enzyme-linked immunosorbent assay and immunoblotting (Western blotting)
- Binds ≥99% to human IgG immobilised on a column