Order Entry
Austria
ContactUsLinkComponent
Methanol ≥99.9% (by GC), ChromAR®, super gradient grade for HPLC, Macron Fine Chemicals™
Methanol ≥99.9% (by GC), ChromAR®, super gradient grade for HPLC, Macron Fine Chemicals™
Catalog # 3041-25
   Safer Human and Environmental Health
CAS Number:  
undefined
Methanol ≥99.9% (by GC), ChromAR®, super gradient grade for HPLC, Macron Fine Chemicals™
Catalog # 3041-25
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.9%
  • Residue after Evaporation:
    Max. 0.0005%
  • Titrable Acid (meq/g):
    Max. 0.0005%
  • Titrable Base (meq/g):
    Max. 0.0002
  • Water (KF):
    Max. 0.05 %
  • at 235 nm:
    Max. 2 mAU
  • at 205 nm:
    Max. 1.00
  • at 210 nm:
    Max. 0.5
  • at 220 nm:
    Max. 0.2
  • at 230 nm:
    Max. 0.1
  • at 240 nm:
    Max. 0.05
  • at 260 nm:
    Max. 0.004

Specification Test Results

About this item

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
 :
 :32.04 g/mol
Merck Index:13,05984
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
Flash Pt:11 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
REACH:01-2119433307-44
 :GHS02
 :GHS06
 :GHS08
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595