Order Entry
Austria
Orders LinkContactUsLinkComponent
Embedding system, Technovit® 9100

Embedding system, Technovit® 9100

Embedding system
Catalog # HKUL64715444
Supplier:   HERAEUS KULZER
Embedding system, Technovit® 9100
Catalog # HKUL64715444
Supplier:  HERAEUS KULZER
Embedding system, Technovit® 9100

Embedding system

Price
543,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Technovit 9100 is a plastic embedding system used in medicine, botany and zoology.

  • Uniform block hardening
  • PMMA block remains transparent
  • Polymerisation below freezing

Designed for mineralised tissues as well as soft tissue with an expanded study spectrum in light microscopy.

The deplasticised sections are suitable for histological overview staining, enzyme chemistry and immunohistological studies, including in situ hybridisation.

Delivery information: Contains a basic solution (1000 ml), PMMA powder (120 g), hardners 1 (8×1 g), hardners 2 (1×10 ml) and a regulator (5 ml).

Specifications
  • Description:
    Embedding system
  • Seq:
    01
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • PMMA powder, 1000 g
    PMMA powder, 1000 g
    HKUL66010251
    Log in for availability
    1 * 1 kg - 524,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Basic solution, 5000 ml
    Basic solution, 5000 ml
    HKUL66006735
    Log in for availability
    1 * 5 L - 550,00 €