Orders LinkContactUsLinkComponent
Process development columns, ToyoScreen® RoboColumn®
Supplier:   TOSOH Bioscience
Process development columns, ToyoScreen® RoboColumn®
Supplier:  TOSOH Bioscience
Certificates
Process development columns, ToyoScreen® RoboColumn®
Process development columns, ToyoScreen® RoboColumn®
32 Options Available Below
Product Details & Documents
Chromatographic columns pre-packed with TOYOPEARL ion exchange, mixed-mode, hydrophobic interaction or affinity media.
  • High throughput parallel chromatography
  • Automated screening of resins and parameters
  • Liquid flow in the columns is driven by positive pressure liquid displacement
These columns can be operated with a robotic liquid handling system, such as the Freedom EVO® from TECAN. This allows automated high-throughput, small-scale bio chromatographic separations of protein samples by running up to eight individual columns simultaneously.
 : Columns are supplied in a row of eight units pre-packed with the TOYOPEARL resin and sealed with two removable silicon cover seals for proper storage.
Product Family Options
Product Information
  • Phase
    Pore size
    Particle Size
    Capacity
    Availability
    Price
  • AF-rProtein A-650F
    100 nm
    45 µm
    200 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 598,00 €
    598,00 €
    1 * 8 ST
       598,00 €
  • AF-rProtein A-650F
    100 nm
    45 µm
    600 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 1 090,00 €
    1 090,00 €
    1 * 8 ST
       1 090,00 €
  • AF-rProtein A HC-650F
    100 nm
    45 µm
    200 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 598,00 €
    598,00 €
    1 * 8 ST
       598,00 €
  • AF-rProtein A HC-650F
    100 nm
    45 µm
    600 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 1 090,00 €
    1 090,00 €
    1 * 8 ST
       1 090,00 €
  • Butyl-600M
    75 nm
    65 µm
    200 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 355,00 €
    355,00 €
    1 * 8 ST
       355,00 €
  • Butyl-600M
    75 nm
    65 µm
    600 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 464,00 €
    464,00 €
    1 * 8 ST
       464,00 €
  • GigaCap CM-650M
    100 nm
    75 µm
    200 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 342,00 €
    342,00 €
    1 * 8 ST
       342,00 €
  • GigaCap CM-650M
    100 nm
    75 µm
    600 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 403,00 €
    403,00 €
    1 * 8 ST
       403,00 €
  • GigaCap DEAE-650M
    100 nm
    75 µm
    200 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 342,00 €
    342,00 €
    1 * 8 ST
       342,00 €
  • GigaCap DEAE-650M
    100 nm
    75 µm
    600 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 403,00 €
    403,00 €
    1 * 8 ST
       403,00 €
  • GigaCap S-650S
    100 nm
    35 µm
    200 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 342,00 €
    342,00 €
    1 * 8 ST
       342,00 €
  • GigaCap S-650S
    100 nm
    35 µm
    600 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 403,00 €
    403,00 €
    1 * 8 ST
       403,00 €
  • GigaCap Q-650S
    100 nm
    75 µm
    200 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 342,00 €
    342,00 €
    1 * 8 ST
       342,00 €
  • GigaCap Q-650S
    100 nm
    75 µm
    600 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 403,00 €
    403,00 €
    1 * 8 ST
       403,00 €
  • GigaCap S-650M
    100 nm
    75 µm
    200 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 342,00 €
    342,00 €
    1 * 8 ST
       342,00 €
  • GigaCap S-650M
    100 nm
    75 µm
    600 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 403,00 €
    403,00 €
    1 * 8 ST
       403,00 €
  • GigaCap Q-650M
    100 nm
    75 µm
    200 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 342,00 €
    342,00 €
    1 * 8 ST
       342,00 €
  • GigaCap Q-650M
    100 nm
    75 µm
    600 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 403,00 €
    403,00 €
    1 * 8 ST
       403,00 €
  • HW-40F
    5 nm
    45 µm
    200 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 342,00 €
    342,00 €
    1 * 8 ST
       342,00 €
  • HW-40F
    5 nm
    45 µm
    600 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 403,00 €
    403,00 €
    1 * 8 ST
       403,00 €
  • MX-Trp-650M
    100 nm
    75 µm
    200 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 355,00 €
    355,00 €
    1 * 8 ST
       355,00 €
  • MX-Trp-650M
    100 nm
    75 µm
    600 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 464,00 €
    464,00 €
    1 * 8 ST
       464,00 €
  • NH2-750F
    100 nm
    45 µm
    200 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 342,00 €
    342,00 €
    1 * 8 ST
       342,00 €
  • NH2-750F
    100 nm
    45 µm
    600 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 403,00 €
    403,00 €
    1 * 8 ST
       403,00 €
  • Phenyl-600M
    75 nm
    65 µm
    200 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 355,00 €
    355,00 €
    1 * 8 ST
       355,00 €
  • Phenyl-600M
    75 nm
    65 µm
    600 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 464,00 €
    464,00 €
    1 * 8 ST
       464,00 €
  • Phenyl-650M
    100 nm
    65 µm
    200 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 355,00 €
    355,00 €
    1 * 8 ST
       355,00 €
  • Phenyl-650M
    100 nm
    65 µm
    600 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 464,00 €
    464,00 €
    1 * 8 ST
       464,00 €
  • PPG-600M
    75 nm
    65 µm
    200 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 355,00 €
    355,00 €
    1 * 8 ST
       355,00 €
  • PPG-600M
    75 nm
    65 µm
    600 µl
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 464,00 €
    464,00 €
    1 * 8 ST
       464,00 €
  • Sulfate-650F
    100 nm
    45 µm
    200 µl
    Log in for availability
    Log in for availability
    1 * 1.600 µl
    1 * 1.600 µl - 342,00 €
    342,00 €
    1 * 1.600 µl
       342,00 €
  • Sulfate-650F
    100 nm
    45 µm
    600 µl
    Log in for availability
    Log in for availability
    1 * 4.800 µl
    1 * 4.800 µl - 403,00 €
    403,00 €
    1 * 4.800 µl
       403,00 €