Order Entry
Austria
ContactUsLinkComponent
Bottles, two necks, rectangular, HDPE
Bottles, two necks, rectangular, HDPE
Catalog # RIXI3-5100-1000-28
Supplier:  RIXIUS
undefined
Bottles, two necks, rectangular, HDPE
Catalog # RIXI3-5100-1000-28
Supplier:  RIXIUS

Specifications

  • Thread:
    OV 28
  • Capacity:
    1000 ml
  • Description:
    Rectangular, two necks
  • W×D×H:
    60×130×255 mm
  • Pk:
    79

Specifications

About this item

Two-neck bottle made up of HDPE, natural.

  • Rigid and shatter-proof
  • Resistant to water, aqueous solutions and most chemicals
  • Excellent water vapour resistance
Ordering information: Screw caps must be ordered seperately.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904