Orders LinkContactUsLinkComponent
Osmolality standards, protein based
Supplier:   REAGECON
Osmolality standards, protein based
Supplier:  REAGECON
Certificates
Osmolality standards, protein based
3 Options Available Below
Product Details & Documents
Osmolality standards provide ease of use when calibrating all types of osmometers.
  • Ready to use and presented in convenient ampoules
  • Extended shelf life and has low uncertainty of measurement
  • Consistency of product-Independent, traceable
  • Consist of 12 ampoules of 5 ml
The Osmolality of the standard is verified using a high performance calibrated, temperature controlled osmometer. The calibration of this instrument is completed using high purity certified Osmolality standards.
 : ISO17025 accreditation for calibration of non automatic weighing machines (INAB ref: 265C). The resulting Balance Certificate of Calibration is issued in accordance with the requirements of ISO/IEC 17025.
Product Family Options
Product Information
  • Osmolality
    Pack type
    Pk Info
    Pk
    Availability
    Price
  • REGERE-POSM-240
    Protein based osmolality standard
    Protein based osmolality standard
    240 mOsm/kg H₂O
    Ampoule
    12× 5 ml
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 174,00 €
    174,00 €
    1 * 1 KIT
       174,00 €
  • REGERE-POSM-280
    Protein based osmolality standard
    Protein based osmolality standard
    280 mOsm/kg H₂O
    Ampoule
    12× 5 ml
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 322,00 €
    322,00 €
    1 * 1 KIT
       322,00 €
  • REGERE-POSM-320
    Protein based osmolality standard
    Protein based osmolality standard
    320 mOsm/kg H₂O
    Ampoule
    12× 5 ml
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 174,00 €
    174,00 €
    1 * 1 KIT
       174,00 €