ContactUsLinkComponent
Peqlab peqGOLD, DNA Ladder, 100 bp Plus
  25-2020
 :  
undefined
Peqlab peqGOLD, DNA Ladder, 100 bp Plus
  25-2020
 :  
  • Pk:
    1 KIT
  • Description:
    DNA ladder, 100 bp plus (concentrated), 500 ng/µl, 1×100 µl, sufficient for 50 lanes
peqGOLD DNA ladder 100 bp plus is ideal for size determination in the range of 100 to 3000 base pairs. There are no unspecific bands besides the fragments described. 1000 and 500 bp bands have a higher DNA content and serve as a size reference.
  • Composed of 14 chromatography-purified individual DNA fragments: 3000, 2000, 1500, 1200, 1000, 900, 800, 700, 600, 500, 400, 300, 200 and 100 base pairs
 : Product comprises of 100 µl (50 µg) DNA ladder and 1 ml 6X loading buffer (10 mM Tris-HCl, pH 7,6; 0,03% bromophenol blue; 0,03% xylene cyanol; 60% glycerol and 60 mM EDTA)
 : The marker has to be dephosphorylated before being used in radioactive assays.