Orders LinkContactUsLinkComponent
Multi-position heating baths
Multi-position heating baths
Supplier:   GESTIGKEIT HARRY
Multi-position heating baths
Supplier:  GESTIGKEIT HARRY
Zertifikate
Multi-position heating baths
Multi-position heating baths
12 Optionen unten verfügbar
Product Details & Documents
Can be universally employed as either an air, fluid or sandbath for heating many of the various receptactles in laboratory use, regardless of size or material, for temperatures ranging between 60 to 600 °C.
  • When in operation the contact thermometer has automatic priority
  • Clearance between baths: 175 mm
  • Type SM without relays
  • Type SMR with relays
  • Voltage: 230 V, 50 to 60 Hz
The bath container is made of high-grade stainless steel with a stainless steel tubular heating element. The upper outer coat is enamelled and the base is made of either plastic or stainless steel.
Type SM features heating elements that can be variably regulated with a performance control between 10 to 100%.
Type SMR features heating elements that are equipped with additional built-in electronic relay switches and an optional output bushing for a contact thermometer.
Without a contact thermometer Type SMR works in exactly the same way as Type SM - with a variable control.
Product Family Options
Produktinformationen
  • Type
    Heating capacity
    Capacity
    W×D×H
    Weight
    Availability
    Price
  • GESTSM1
    Heating bath with one heating unit
    Heating bath with one heating unit
    SM 1
    500 W
    1 L
    175×245×215 mm
    2,8 kg
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 ST
    1 * 1 ST
  • GESTSM2
    Heating bath with two heating units
    Heating bath with two heating units
    SM 2
    1000 W
    2 × 1 L
    350×245×215 mm
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 ST
    1 * 1 ST
  • GESTSM3
    Heating bath with three heating units
    Heating bath with three heating units
    SM 3
    1500 W
    3 × 1 L
    525×245×215 mm
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 ST
    1 * 1 ST
  • GESTSM4
    Heating bath with four heating units
    Heating bath with four heating units
    SM 4
    2000 W
    4 × 1 L
    700×245×215 mm
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 ST
    1 * 1 ST - 1 200,00 €
    1 200,00 €
    1 * 1 ST
       1 200,00 €
  • GESTSM5
    Heating bath with five heating units
    Heating bath with five heating units
    SM 5
    2500 W
    5 × 1 L
    875×245×215 mm
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 ST
    1 * 1 ST
  • GESTSM6
    Heating bath with six heating units
    Heating bath with six heating units
    SM 6
    3000 W
    6 × 1 L
    1050×245×215 mm
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 ST
    1 * 1 ST
  • GESTSMR1
    Heating bath with one heating unit and relay
    Heating bath with one heating unit and relay
    SMR 1
    500 W
    1 L
    175×245×215 mm
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 ST
    1 * 1 ST
  • GESTSMR2
    Heating bath with two heating units and relay
    Heating bath with two heating units and relay
    SMR 2
    1000 W
    2 × 1 L
    350×245×215 mm
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 ST
    1 * 1 ST
  • GESTSMR3
    Heating bath with three heating units and relay
    Heating bath with three heating units and relay
    SMR 3
    1500 W
    3 × 1 L
    525×245×215 mm
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 ST
    1 * 1 ST
  • GESTSMR4
    Heating bath with four heating units and relay
    Heating bath with four heating units and relay
    SMR 4
    2000 W
    4 × 1 L
    700×245×215 mm
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 ST
    1 * 1 ST
  • GESTSMR5
    Heating bath with five heating units and relay
    Heating bath with five heating units and relay
    SMR 5
    2500 W
    5 × 1 L
    875×245×215 mm
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 ST
    1 * 1 ST
  • GESTSMR6
    Heating bath with six heating units and relay
    Heating bath with six heating units and relay
    SMR 6
    3000 W
    6 × 1 L
    1050×245×215 mm
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 ST
    1 * 1 ST