Wash bottles, integral, wide neck, multilingual, Azlon®
Supplier: SciLabware
Certificates
About this item
Multilingual, wide neck wash bottles with coloured caps.
- Indelibly printed in English, French, German and Spanish
- Fitted with colour coded one-piece PP closure that maintains it's shape in use
- Vented cap with PTFE membrane allows venting and prevents pressure build up
- Designed with integral spout to prevent contamination
- Good chemical resistance
This round wash bottle is moulded with an integral spout so you can say goodbye to contamination caused by the inner tube touching the bench top during re-filling. Choose from five solvent labels all including the new Globally Harmonised Symbols, NFPA diamond, risk phrases and safety information.
Caution: NOTE: For dispensing use only. Not for liquid storage.
Frequently Bought Together