Order Entry
Austria
ContactUsLinkComponent
VWR® PerfectBlue Maxi ExW, Wide format gel system
VWR® PerfectBlue Maxi ExW, Wide format gel system
Catalog # 700-0910
Supplier:  VWR Collection
undefined
VWR® PerfectBlue Maxi ExW, Wide format gel system
Catalog # 700-0910
Supplier:  VWR Collection

Specifications

  • Description:
    PerfectBlue gel system Maxi ExW
  • Model:
    Maxi ExW
  • Gel volume:
    290 ml
  • Buffer volume:
    ~3000 ml
  • Run time:
    ≥60 h
  • Gel size W×L:
    230×250 mm
  • Sample capacity:
    25 − 500
  • Unit W×L×H:
    300×370×110 mm
  • Pk:
    1

Specifications

About this item

This 'extra wide' Maxi tank was developed especially for large sample numbers and rapid separation times. Accommodating up to 10 microtitre combs, the Maxi ExW can be used to separate up to 500 samples and facilitates error-free loading with multichannel pipettes. By optimising the position of the embedded platinum electrode and the use of the three-point levelling platform, with in-built bubble level, the Maxi ExW provides highly reproducible and dependable sample separation, free from 'smiling' effects.

The tray has 10 comb positions accomodating two different comb thicknesses. The tray is designed for use with sturdy 'end gates' for in-tray gel pouring and economical 'tray partitions' that enable smaller gels to be poured in the same tray - saving time and money.

* Based on an agarose gel of approximately 5 mm thickness.

Delivery: Supplied with 4 combs (1,5 mm thickness, 2×25 and 2×50 teeth), lid, gel tray, 3-point levelling platform, end gates, tank and cables.

Combs

Product Information