Order Entry
Austria
ContactUsLinkComponent
 
VWR® PerfectBlue Maxi S Plus, Gel system
  700-0876
undefined
VWR® PerfectBlue Maxi S Plus, Gel system
  700-0876

 

  • Description:
    PerfectBlue gel system Maxi S Plus
  • Model:
    Maxi S Plus
  • Gel volume:
    200 ml
  • Buffer volume:
    ~1800 ml
  • Run time:
    ≥60 h
  • Gel size W×L:
    150×250 mm
  • Sample capacity:
    10 − 160
  • Unit W×L×H:
    230×375×105 mm
  • Pk:
    1

 

 

The smallest of the Maxi series, the Maxi S Plus is suitable for high quality sample separation of up to 160 samples per run. With this flexible system, the gel size can be split into two, three or four sections by using different comb positions, plus the gel size can be reduced using the wall comb or the casting dam system.

The system is supplied with three combs but others are available – either standard combs, or microtitre combs which are ideal for use with multichannel pipettes. Use of the system end gates ensure that gel pouring is leak-free and easy. The levelling table keeps the resulting gel flat, maximising the quality of the run.

* Based on an agarose gel of approximately 5 mm thickness.

 : Supplied with 3 combs (1,5 mm thickness, 10, 17, 34 teeth), lid, gel tray, 3-point levelling platform, tank, cables and end gates.

Combs