

(S)-(-)-Nicotine for synthesis, Sigma-Aldrich®
: Merck
:54-11-5
:
:C₁₀H₁₄N₂
:MFCD00006369
:12,06611
:247 °C (1013 hPa)
:–79 °C
:1.01 g/cm³ (20 °C)
:>100 °C
:200-193-3
:1654
:6.1,II
:M-Clarity™ quality level = MQ200
:162.23 g/mol
:GHS06
:GHS09
:Ambient
- Solubility:(20 °C) soluble
- Melting Point:-79 °C
- Molar Mass:162.23 g/mol
- Boiling Point:247 °C (1013 hPa)
- Vapor Pressure:0.05 hPa (25 °C)
- Flash Point:101 °C
- Explosion Limit:0.7 - 4 %(V)
- Density:1.01 g/cm3 (20 °C)
- pH:10.2 (8.1 g/l, H2O, 20 °C)
- Ignition Point:240 °C
- Synonyms:3-(1-Methyl-2-pyrrolidinyl)pyridine, (S)-(-)-Nicotine
- Hill Formula:C10H14N2
Frequently Bought Together