Order Entry
Austria
ContactUsLinkComponent
 
Human Recombinant M-CSF (from E. coli)
 
undefined
Human Recombinant M-CSF (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: NFS-60 cell proliferation, ED50 ≤10 ng/ml, bioactivity ≥1,0×10⁵ units/mg
  • Gene ID:
    P09603
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    MGI-IM|CSF-1
  • UniProtKB:
    P09603
  • Protein/peptide name:
    M-CSF
  • Purity:
    ≥95%
  • Molecular weight:
    18,5/37,1 kDa
  • Sequence:
    MEEVSEYCSH MIGSGHLQSL QRLIDSQMET SCQITFEFVD QEQLKDPVCY LKKAFLLVQD IMEDTMRFRD NTPNAIAIVQ LQELSLRLKS CFTKDYEEHD KACVRTFYET PLQLLEKVKN VFNETKNLLD KDWNIFSKNC NNSFAECSSQ GHERQSEGS
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, 50 mM sodium chloride, pH 8,0
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

 

Frequently Bought Together