4-Acetylpyridine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1122-54-9
:
Formula:C₇H₇NO
MDL Number:MFCD00006433
:121.14 g/mol
Boiling Pt:212 °C (1013 hPa)
Melting Pt:14…16 °C
Density:1.1009 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:214-350-9
Frequently Bought Together