
2-Pyridinethiol 98%
:2637-34-5
:73018-10-7
:C₅H₅NS
:MFCD00006285
:111.17 g/mol
:259 °C (1013 hPa)
:126…130 °C
:125 °C
:Refrigerator
:220-131-9
:GHS07
- Appearance:Yellow to brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:126 to 130 °C
- Titration Iodimetric:≥97.5 %
Frequently Bought Together