Human Recombinant Omentin (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Omentin is an adipokine that is produced and secreted by the small intestine, visceral adipose tissue, perivascular adipose tissue, and epicardial adipose tissue. Omentin enhances insulin-stimulated glucose uptake in adipocytes and is a link between obesity and Type 2 Diabetes. Omentin also functions as a vasodilator and plays a protective role during coronary atherosclerosis and hypertension.
- High Quality
- Low Endotoxin
- Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤5
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q8WWA0
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:Intelectin-1|ITLN-1
- Protein/peptide name:Omentin
- Purity:≥90%
- Molecular weight:35 kDa
- Sequence:MNQLSFLLFL IATTRGWSTD EANTYFKEWT CSSSPSLPRS CKEIKDECPS AFDGLYFLRT ENGVIYQTFC DMTSGGGGWT LVASVHENDM RGKCTVGDRW SSQQGSKADY PEGDGNWANY NTFGSAEAAT SDDYKNPGYY DIQAKDLGIW HVPNKSPMQH WRNSSLLRYR TDTGFLQTLG HNLFGIYQKY PVKYGEGKCW TDNGPVIPVV YDFGDAQKTA SYYSPYGQRE FTAGFVQFRV FNNERAANAL CAGMRVTGCN TEHHCIGGGG YFPEASPQQC GDFSGFDWSG YGTHVGYSSS REITEAAVLL FYR
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate and 5:1 mannitol to protein, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together