Nicotinamide, Millipore®
481907-100
:
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
:
:12,06574
:334 °C (1013 hPa)
:127…128 °C
:1.4463 g/cm³ (20 °C)
:182 °C
:122.13 g/mol
:Ambient
:202-713-4
:GHS07
:98-92-0
:C₆H₆N₂O
:MFCD00006395