Order Entry
Austria
ContactUsLinkComponent
Pyridine-2,6-dicarboxylic acid for synthesis, Sigma-Aldrich®
Irritant
Pyridine-2,6-dicarboxylic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
Pyridine-2,6-dicarboxylic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    5 g/l (20 °C)
  • Melting Point:
    224 - 255 °C
  • Molar Mass:
    167.12 g/mol
  • Bulk Density:
    700 kg/m3
  • Vapor Pressure:
     - 0.01 hPa (20 °C)
  • Flash Point:
    188 °C
  • pH:
    2.0 (5 g/l, H2O, 20 °C)
  • Ignition Point:
    620 °C

Specification Test Results

Related Information

Frequently Bought Together