Order Entry
Austria
ContactUsLinkComponent
Pyridine-2,6-dicarboxylic acid 99%
Irritant
Pyridine-2,6-dicarboxylic acid 99%
Certificates
undefined
Irritant
Pyridine-2,6-dicarboxylic acid 99%

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.3 % (K.F.)
  • Titration with NaOH:
    98.5 to 101.5 %

Specification Test Results

Related Information

Frequently Bought Together