Order Entry
Austria
ContactUsLinkComponent
Isonicotinic acid for synthesis, Sigma-Aldrich®
Irritant
Isonicotinic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
Isonicotinic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Hill Formula:
    C₆H₅NO₂
  • Molar Mass:
    123.11 g/mol
  • Boiling point:
    260 °C (20 hPa)
  • Melting Point:
    315 - 319 °C
  • pH value:
    3 - 4 (6 g/l, H₂O, 20 °C) (saturated solution)
  • Bulk density:
    500 kg/m³
  • Solubility:
    6 g/l (20 °C)
  • LD 50 oral:
    LD50 Rat 5000 mg/kg
  • Storage class:
    10 - 13 Other liquids and solids
  • WGK:
    WGK 1 slightly water endangering
  • R Phrase:
    R 36
  • S Phrase:
    S 22-24-26

Specification Test Results

Related Information

Frequently Bought Together