
Methyl nicotinate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:93-60-7
:
Formula:C₇H₇NO₂
MDL Number:MFCD00006388
:137.14 g/mol
Merck Index:12,06174
Boiling Pt:209 °C (1013 hPa)
Melting Pt:39 °C
Density:1.0048 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:95 °C
EINECS:202-261-8
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Solubility:48 g/l (20 °C)
- Melting Point:39 °C
- Molar Mass:137.14 g/mol
- Bulk Density:600 kg/m3
- Boiling Point:209 °C (1013 hPa)
- Vapor Pressure:0.4 hPa (25 °C)
- Flash Point:95 °C
Specification Test Results
Related Information
Frequently Bought Together