Order Entry
Austria
ContactUsLinkComponent
6-Chloronicotinic acid
Irritant
6-Chloronicotinic acid
Catalog # EHERC11452000
Supplier:  EHRENSTORFER
CAS Number:  
Irritant
6-Chloronicotinic acid
Catalog # EHERC11452000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :157.56 g/mol
Boiling Pt:284 °C (1013 hPa)
Melting Pt:188…189 °C
EINECS:226-201-5
 :GHS07
Storage Temperature:Ambient
CAS Number:5326-23-8
Formula:C₆H₄ClNO₂
MDL Number:MFCD00006241