Order Entry
Austria
ContactUsLinkComponent
Calibration standard, NexION KED setup solution
Calibration standard, NexION KED setup solution
Catalog # PERCN8145289
Supplier:  PerkinElmer
CAS Number:  
Calibration standard, NexION KED setup solution
Catalog # PERCN8145289
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Pk:
    500 mL
  • Description:
    NexION KED setup solution in 1% HNO3
  • Contents:
    Be, Ce, Fe, In, Li, Mg, Pb, U

Specifications

About this item