About this item
Galectin-1 belongs to the lectin family of carbohydrate binding proteins and binds glycans as both a monomer and a homodimer. Galectin-1 is produced in peripheral lymphoid organs and inflammatory sites. Galectin-1 plays important roles in acute and chronic inflammatory processes, cell growth, cell proliferation, and induces apoptosis of activated T cells. Galectin-1 also modulates cytokine secretion and inhibits pro-inflammatory cytokine production.
- High quality
- Low endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P09382
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:S-LAC lectin-I|L-14|Beta-galactoside-binding lectin L-14-I|galaptin
- UniProtKB:P09382
- Protein/peptide name:Galectin-1
- Purity:≥95%
- Molecular weight:14,7 kDa
- Sequence:MACGLVASNL NLKPGECLRV RGEVAPDAKS FVLNLGKDSN NLCLHFNPRF NAHGDANTIV CNSKDGGAWG TEQREAVFPF QPGSVAEVCI TFDQANLTVK LPDGYEFKFP NRLNLEAINY MAADGDFKIK CVAFD
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT