Rat Recombinant EGF (from E. coli)
Epidermal growth factor (EGF) is a growth factor that stimulates the proliferation, differentiation, and survival of epithelial and epidermal cells. EGF contains three intramolecular disulfide bonds and binds in high affinity to the epidermal growth factor receptor (EGFR).
- High Quality
- Low Endotoxin
- Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
: For research use only.
Epidermal growth factor (EGF) is a growth factor that stimulates the proliferation, differentiation, and survival of epithelial and epidermal cells. EGF contains three intramolecular disulfide bonds and binds in high affinity to the epidermal growth factor receptor (EGFR).
- High Quality
- Low Endotoxin
- Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
: For research use only.
- Catalog No:
- SHBT300-05-20UG
- SHBT300-05-100UG
- SHBT300-05-250UG
- SHBT300-05-500UG
- SHBT300-05-1MG
- Protein/peptide name:
- EGF
- EGF
- EGF
- EGF
- EGF
- Protein synonyms:
- Urogastrone|URG
- Urogastrone|URG
- Urogastrone|URG
- Urogastrone|URG
- Urogastrone|URG
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein function:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Species:
- Rat
- Rat
- Rat
- Rat
- Rat
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- P07522
- P07522
- P07522
- P07522
- P07522
- Sequence:
- MNSNTGCPPS YDGYCLNGGV CMYVESVDRY VCNCVIGYIG ERCQHRDLRW WKLR
- MNSNTGCPPS YDGYCLNGGV CMYVESVDRY VCNCVIGYIG ERCQHRDLRW WKLR
- MNSNTGCPPS YDGYCLNGGV CMYVESVDRY VCNCVIGYIG ERCQHRDLRW WKLR
- MNSNTGCPPS YDGYCLNGGV CMYVESVDRY VCNCVIGYIG ERCQHRDLRW WKLR
- MNSNTGCPPS YDGYCLNGGV CMYVESVDRY VCNCVIGYIG ERCQHRDLRW WKLR
- Protease-free:
- N
- N
- N
- N
- N
- Animal-free:
- N
- N
- N
- N
- N
- Nuclease-free:
- N
- N
- N
- N
- N
- Carrier-free:
- Y
- Y
- Y
- Y
- Y
- Endotoxin-free:
- N
- N
- N
- N
- N
- Endotoxin level:
- Low
- Low
- Low
- Low
- Low
- Endotoxin content:
- Endotoxin LAL (EU/µg) ≤1
- Endotoxin LAL (EU/µg) ≤1
- Endotoxin LAL (EU/µg) ≤1
- Endotoxin LAL (EU/µg) ≤1
- Endotoxin LAL (EU/µg) ≤1
- Biological activity:
- Bioactivity assay: BALB/c 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
- Bioactivity assay: BALB/c 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
- Bioactivity assay: BALB/c 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
- Bioactivity assay: BALB/c 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
- Bioactivity assay: BALB/c 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
- Purity:
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- Grade:
- RUO
- RUO
- RUO
- RUO
- RUO
- Formulation:
- 10 mM sodium phosphate, pH 7,5
- 10 mM sodium phosphate, pH 7,5
- 10 mM sodium phosphate, pH 7,5
- 10 mM sodium phosphate, pH 7,5
- 10 mM sodium phosphate, pH 7,5
- Molecular weight:
- 6,3 kDa
- 6,3 kDa
- 6,3 kDa
- 6,3 kDa
- 6,3 kDa
- Reconstitution instructions:
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Storage conditions:
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- Shipping temperature:
- RT
- RT
- RT
- RT
- RT
- Tested applications:
- Bioassay
- Bioassay
- Bioassay
- Bioassay
- Bioassay
Product Family Options
- Pack typePk
Specifications
Cat. No.SHBT300-05-20UGProtein/peptide nameEGFProtein synonymsUrogastrone|URGProtein/peptide typeRecombinantProtein functionCytokineSpeciesRatSourceE. coliConjugationUnconjugatedGene IDP07522SequenceMNSNTGCPPS YDGYCLNGGV CMYVESVDRY VCNCVIGYIG ERCQHRDLRW WKLRProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤1Biological activityBioactivity assay: BALB/c 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mgPurity≥95%GradeRUOFormulation10 mM sodium phosphate, pH 7,5Molecular weight6,3 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassaySpecifications
Cat. No.SHBT300-05-100UGProtein/peptide nameEGFProtein synonymsUrogastrone|URGProtein/peptide typeRecombinantProtein functionCytokineSpeciesRatSourceE. coliConjugationUnconjugatedGene IDP07522SequenceMNSNTGCPPS YDGYCLNGGV CMYVESVDRY VCNCVIGYIG ERCQHRDLRW WKLRProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤1Biological activityBioactivity assay: BALB/c 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mgPurity≥95%GradeRUOFormulation10 mM sodium phosphate, pH 7,5Molecular weight6,3 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassaySpecifications
Cat. No.SHBT300-05-250UGProtein/peptide nameEGFProtein synonymsUrogastrone|URGProtein/peptide typeRecombinantProtein functionCytokineSpeciesRatSourceE. coliConjugationUnconjugatedGene IDP07522SequenceMNSNTGCPPS YDGYCLNGGV CMYVESVDRY VCNCVIGYIG ERCQHRDLRW WKLRProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤1Biological activityBioactivity assay: BALB/c 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mgPurity≥95%GradeRUOFormulation10 mM sodium phosphate, pH 7,5Molecular weight6,3 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassaySpecifications
Cat. No.SHBT300-05-500UGProtein/peptide nameEGFProtein synonymsUrogastrone|URGProtein/peptide typeRecombinantProtein functionCytokineSpeciesRatSourceE. coliConjugationUnconjugatedGene IDP07522SequenceMNSNTGCPPS YDGYCLNGGV CMYVESVDRY VCNCVIGYIG ERCQHRDLRW WKLRProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤1Biological activityBioactivity assay: BALB/c 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mgPurity≥95%GradeRUOFormulation10 mM sodium phosphate, pH 7,5Molecular weight6,3 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassaySpecifications
Cat. No.SHBT300-05-1MGProtein/peptide nameEGFProtein synonymsUrogastrone|URGProtein/peptide typeRecombinantProtein functionCytokineSpeciesRatSourceE. coliConjugationUnconjugatedGene IDP07522SequenceMNSNTGCPPS YDGYCLNGGV CMYVESVDRY VCNCVIGYIG ERCQHRDLRW WKLRProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤1Biological activityBioactivity assay: BALB/c 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mgPurity≥95%GradeRUOFormulation10 mM sodium phosphate, pH 7,5Molecular weight6,3 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



