ContactUsLinkComponent
Ringers solution ¹/₄ strength tablets, NutriSelect® Basic, Millipore®
Ringers solution ¹/₄ strength tablets, NutriSelect® Basic, Millipore®
Catalog # SIAL96724-100TAB
Supplier:  Merck
CAS Number:  
Ringers solution ¹/₄ strength tablets, NutriSelect® Basic, Millipore®
Catalog # SIAL96724-100TAB
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    100 Tab.
  • Description:
    Ringers solution 1/4 strength tablets
Specifications
About this item
Ringers solution maintains the osmotic balance for bacteria.
  • One tablet is sufficient for 500 ml
Used for the preparation of suspension of samples and for use as a diluent in dilution techniques for enumeration. Also used in the microbial control of apparatus by the rinse and swab method.