L(+)-Ascorbic acid ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99+% CAS No.: 50-81-7 Molecular Formula: C6H8O6 Formula Weight: 176.12 Melting Point: 190-193° dec. Beilstein Reference: 84272 Merck Reference: 14,830 Optical Rotation: +21° (c=10 in water) MDL No.: MFCD00064328
Synonyms: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
- CAS Number:50-81-7
- Formula:C₆H₈O₆
- MDL Number:MFCD00064328
- Molecular Weight:176.13 g/mol
- Merck Index:14,830
- Boiling Pt:553 °C (1013 hPa)
- Melting Pt:190…193 °C (dec.)
- Density:1.65
- Storage Temperature:Ambient
- EINECS:200-066-2
- REACH:exempt
3 Options Available Below
Product Details & Documents
Purity: 99+% CAS No.: 50-81-7 Molecular Formula: C6H8O6 Formula Weight: 176.12 Melting Point: 190-193° dec. Beilstein Reference: 84272 Merck Reference: 14,830 Optical Rotation: +21° (c=10 in water) MDL No.: MFCD00064328
Synonyms: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
- CAS Number:50-81-7
- Formula:C₆H₈O₆
- MDL Number:MFCD00064328
- Molecular Weight:176.13 g/mol
- Merck Index:14,830
- Boiling Pt:553 °C (1013 hPa)
- Melting Pt:190…193 °C (dec.)
- Density:1.65
- Storage Temperature:Ambient
- EINECS:200-066-2
- REACH:exempt
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



