Order Entry
Austria
ContactUsLinkComponent
Iron(III) oxide, anhydrous ≥97%, TECHNICAL
Irritant
Iron(III) oxide, anhydrous ≥97%, TECHNICAL
Catalog # 24189.468
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Iron(III) oxide, anhydrous ≥97%, TECHNICAL
Catalog # 24189.468
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bucket
  • Pk:
    25 kg
  • Seq:
    4283

Specifications

Specification Test Results

  • Assay:
    Min. 97 %

Specification Test Results

About this item

 :
 :159.69 g/mol
Merck Index:13,04058
Melting Pt:1538 °C
Density:5.24 g/cm³ (20 °C)
EINECS:215-168-2
 :GHS07
CAS Number:1309-37-1
Formula:Fe₂O₃
MDL Number:MFCD00011008