
Iron(III) oxide 99.999% (trace metals basis) -100 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1309-37-1
:
Formula:Fe₂O₃
MDL Number:MFCD00011008
:159.69 g/mol
Merck Index:13,04058
Melting Pt:1538 °C (dec.)
Density:5.24 g/cm³
EINECS:215-168-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Red-brown to rust brown Powder
- Total trace metal impurities:≤0.001 %
Specification Test Results
Frequently Bought Together