
Platinum(IV) iodide ≥99.95% (metals basis) Pt ≥ 27.3%, Premion®
Catalog # 40401.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
About this item
CAS Number:7790-46-7
:
Formula:I₄Pt
MDL Number:MFCD00016292
:702.7 g/mol
Merck Index:12,07685
Melting Pt:130 °C (dec.)
Density:6.06
Storage Temperature:Ambient
EINECS:232-207-9
UN:3260
ADR:8,III
:GHS05