Rhodium(III) chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10049-07-7
:
Formula:Cl₃Rh
MDL Number:MFCD00011204
:209.26 g/mol
Merck Index:12,08355
Density:5.38 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:233-165-4
Frequently Bought Together