Order Entry
Austria
ContactUsLinkComponent
Nickel(II) chloride for synthesis, Sigma-Aldrich®
Nickel(II) chloride for synthesis, Sigma-Aldrich®
Catalog # 8.06722.0100
Supplier:  Merck
CAS Number:  
Nickel(II) chloride for synthesis, Sigma-Aldrich®
Catalog # 8.06722.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    1009 °C
  • Molar Mass:
    129.6 g/mol
  • Vapor Pressure:
    1.33 hPa (671 °C)
  • Density:
    3.55 g/cm3
  • pH:
    4 (500 g/l, H2O, 20 °C)

Specification Test Results

About this item

 :
 :129.6 g/mol
Merck Index:12,06588
Melting Pt:1009 °C
Density:3.55 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-743-0
UN:3288
ADR:6.1,III
 :M-Clarity™ quality level = MQ200
 :GHS06
 :GHS08
 :GHS09
CAS Number:7718-54-9
Formula:Cl₂Ni
MDL Number:MFCD00011142